1,3-Dimethyl-1H-pyrazol-5-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL722340-1GEA 51.4 EUR
SIAL722340-1G
1,3-Dimethyl-1H-pyrazol-5-carbonsäure, Sigma-Aldrich®
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₈N₂O₂
Molekulargewicht: 140,1400 g/mol
MDL Number: MFCD00159636
CAS-Nummer: 5744-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR