Aceton-[13C3] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ quality level = MQ200
SIAL491667-250MGEA
661
EUR
SIAL491667-250MG
Aceton-[13C3] (99% ¹³C ), Sigma-Aldrich®
Aceton-[13C3]
Formel:
C₃H₆O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00083865 CAS-Nummer: 93628-01-4 UN-Nummer: 1090 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...