2-Chlorethyl-methylsulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL242632-5GEA 76 EUR
SIAL242632-5G
2-Chlorethyl-methylsulfid, Sigma-Aldrich®
2-Chlorethyl-methylsulfid
Formel: C₃H₇ClS
MDL Number: MFCD00000978
CAS-Nummer: 542-81-4
EINECS: 208-828-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR