Flufenoxuron
Lieferant: EHRENSTORFER GMBH
EHERC13712000EA
115
EUR
EHERC13712000
Flufenoxuron
Flufenoxuron
Formel:
C₂₁H₁₁ClF₆N₂O₃ |
MDL Number:
MFCD00274597 CAS-Nummer: 101463-69-8 EINECS: 417-680-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...