Flufenoxuron, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

31594-250MGEA 158 EUR
31594-250MG
Flufenoxuron, Supelco®
Flufenoxuron
Formel: C₂₁H₁₁ClF₆N₂O₃
MDL Number: MFCD00274597
CAS-Nummer: 101463-69-8
EINECS: 417-680-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR