3-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL527548-1GEA
34
EUR
SIAL527548-1G
3-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ Molekulargewicht: 276,136 g/mol |
MDL Number:
MFCD03093896 CAS-Nummer: 269410-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...