3-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL574694-1GEA
30.5
EUR
SIAL574694-1G
3-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
3-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molekulargewicht: 248,083 g/mol |
MDL Number:
MFCD03411930 CAS-Nummer: 269409-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...