4-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL513490-5GEA
28.1
EUR
SIAL513490-5G
4-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
4-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molekulargewicht: 248,08 g/mol |
MDL Number:
MFCD01863710 CAS-Nummer: 180516-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...