3-(2,5-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%

H52504.03EA 201 EUR
ALFAH52504.03 ALFAH52504.MD
3-(2,5-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 957061-00-6
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027263
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR