4-(2,3-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
H53189.03EA 109 EUR
ALFAH53189.03 ALFAH53189.MD
4-(2,3-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-36-6
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027210