4-(2,4-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
H53356.03EA 201 EUR
ALFAH53356.03 ALFAH53356.MD
4-(2,4-Dimethylphenylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-38-8
Molecular Formula: C15H16BNO3
Formula Weight: 269.11
MDL No.: MFCD09027212