3-Aminophenylboronsäure Hemisulfat

Lieferant: Molekula

MOLE44219424-5GEA 167 EUR
MOLE44219424-5G
3-Aminophenylboronsäure Hemisulfat
3-Aminophenylboronsäure Hemisulfat
Formel: C₁₂H₁₈B₂N₂O₈S
Molekulargewicht: 371,97 g/mol
MDL Number: MFCD00013111
CAS-Nummer: 66472-86-4
EINECS: 266-376-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR