2-Chloro-4-[methyl(4-nitrophenyl)carbamoyl]benzeneboronic acid, 97%

H55551.03EA 343 EUR
ALFAH55551.03 ALFAH55551.MD
2-Chloro-4-[methyl(4-nitrophenyl)carbamoyl]benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClN2O5
Formula Weight: 334.52
MDL No.: MFCD22125152
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR