2-Chloro-4-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54047.03EA 246 EUR
ALFAH54047.03 ALFAH54047.MD
2-Chloro-4-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125038
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR