3-Chloro-4-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54701.MDEA 47.6 EUR
ALFAH54701.MD ALFAH54701.03
3-Chloro-4-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265325
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR