3-Chloro-4-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54028.MDEA 35.8 EUR
ALFAH54028.MD ALFAH54028.03
3-Chloro-4-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265162
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR