2-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, 98%
H52963.03EA 115 EUR
ALFAH52963.03 ALFAH52963.MD
2-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-94-6
Molecular Formula: C9H10BClO4
Formula Weight: 228.44
Melting Point: 115-118°
MDL No.: MFCD08689531