3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, 98%

H52896.03EA 111 EUR
ALFAH52896.03 ALFAH52896.MD
3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 850568-11-5
Molecular Formula: C9H10BClO4
Formula Weight: 228.44
Melting Point: 122-126°
MDL No.: MFCD06659875
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR