3-Chloro-4-(methoxycarbonyl)benzeneboronic acid, 97%

H53271.MDEA 16.1 EUR
ALFAH53271.MD ALFAH53271.03
3-Chloro-4-(methoxycarbonyl)benzeneboronic acid, 97%
Purity: 97%
CAS No.: 603122-82-3
Molecular Formula: C8H8BClO4
Formula Weight: 214.41
Melting Point: 174-178°
MDL No.: MFCD06659874
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR