4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 98%

H52854.03EA 77.2 EUR
ALFAH52854.03 ALFAH52854.MD
4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-45-1
Molecular Formula: C8H8BClO4
Formula Weight: 214.41
Melting Point: 136-138°
MDL No.: MFCD06801690
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR