3-Carboxy-4-chlorobenzeneboronic acid, 98%

H52764.03EA 81 EUR
ALFAH52764.03 ALFAH52764.MD
3-Carboxy-4-chlorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-32-2
Molecular Formula: C7H6BClO4
Formula Weight: 200.39
Melting Point: 223-225°
MDL No.: MFCD06801689
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR