2-Chloro-5-(5-fluoro-2-methylphenylcarbamoyl)benzeneboronic acid, 97%

H55813.03EA 191 EUR
ALFAH55813.03 ALFAH55813.MD
2-Chloro-5-(5-fluoro-2-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040328
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR