2-Amino-4-chlor-6-nitrophenol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL530387-5GEA
41.7
EUR
SIAL530387-5G
SIAL530387-25G
2-Amino-4-chlor-6-nitrophenol, Sigma-Aldrich®
2-Amino-4-chlor-6-nitrophenol
Formel:
C₆H₅ClN₂O₃ Molekulargewicht: 188,57 g/mol |
MDL Number:
MFCD00035925 CAS-Nummer: 6358-08-3 EINECS: 228-761-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...