Asulam

Lieferant: EHRENSTORFER GMBH

EHERC10310000EA 47.2 EUR
EHERC10310000
Asulam
Asulam
Formel: C₈H₁₀N₂O₄S
MDL Number: MFCD00055534
CAS-Nummer: 3337-71-1
EINECS: 222-077-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR