4-(2,4-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

H55803.03EA 286 EUR
ALFAH55803.03 ALFAH55803.MD
4-(2,4-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040327
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR