4-(2,6-Dimethylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%

H54603.03EA 343 EUR
ALFAH54603.03 ALFAH54603.MD
4-(2,6-Dimethylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040228
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR