2-Fluoro-5-(4-methylphenylcarbamoyl)benzeneboronic acid, 97%

H54324.MDEA 85.7 EUR
ALFAH54324.MD ALFAH54324.03
2-Fluoro-5-(4-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20265241
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR