4-(Methylsulfonyl)phenol

Lieferant: Thermo Scientific

MAYBJFD01524.1EA 0 EUR
MAYBJFD01524.1 CHMPJFD01524.250
4-(Methylsulfonyl)phenol
4-(Methylsulfonyl)phenol
Formel: C₇H₈O₃S
MDL Number: MFCD00047806
CAS-Nummer: 14763-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR