p-Toluolsulfonylfluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL117455-10GEA 44.5 EUR
SIAL117455-10G
p-Toluolsulfonylfluorid, Sigma-Aldrich®
p-Toluolsulfonylfluorid
Formel: C₇H₇FO₂S
Molekulargewicht: 174,19 g/mol
MDL Number: MFCD00007421
CAS-Nummer: 455-16-3
EINECS: 207-238-6
UN-Nummer: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR