2-Methoxy-3-(trifluormethyl)benzylamin 97%, JRD
|
Gefahr
|
H32993.03EA
167
EUR
H32993.03
H32993.MD
2-Methoxy-3-(trifluormethyl)benzylamin 97%, JRD
2-Methoxy-3-(trifluormethyl)benzylamin
Formel:
C₉H₁₀F₃NO Molekulargewicht: 205,18 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832318 CAS-Nummer: 1017778-87-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...