2-Biphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL542202-5GEA 132 EUR
SIAL542202-5G
2-Biphenylboronsäure, Sigma-Aldrich®
2-Biphenylboronsäure
Formel: C₁₂H₁₁BO₂
Molekulargewicht: 198,03
MDL Number: MFCD00136929
CAS-Nummer: 4688-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR