5-Benzyl(phenyl)carbamoyl-2-chlorobenzeneboronic acid, 97%

H54238.MDEA 61.3 EUR
ALFAH54238.MD ALFAH54238.03
5-Benzyl(phenyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C20H17BClNO3
Formula Weight: 365.62
MDL No.: MFCD22125052
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR