3-Chloro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
H54377.03EA 214 EUR
ALFAH54377.03 ALFAH54377.MD
3-Chloro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265254