2-Chloro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H55730.MDEA 53.6 EUR
ALFAH55730.MD ALFAH55730.03
2-Chloro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125170
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR