2-Chloro-4-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H55157.03EA 172 EUR
ALFAH55157.03 ALFAH55157.MD
2-Chloro-4-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040292
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR