3-Chloro-4-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Lieferant: Thermo Scientific
H54164.03EA 156 EUR
ALFAH54164.03 ALFAH54164.MD
3-Chloro-4-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040144