4-Chloro-3-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H54518.MDEA 47.6 EUR
ALFAH54518.MD ALFAH54518.03
4-Chloro-3-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040211
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR