4-[Benzyl(ethyl)carbamoyl]-3-fluorobenzeneboronic acid, 97%

H54468.MDEA 85.7 EUR
ALFAH54468.MD ALFAH54468.03
4-[Benzyl(ethyl)carbamoyl]-3-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C16H17BFNO3
Formula Weight: 301.12
MDL No.: MFCD20040194
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR