3-Fluoro-4-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H54544.MDEA 85.7 EUR
ALFAH54544.MD ALFAH54544.03
3-Fluoro-4-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040216
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR