3-Benzylcarbamoyl-4-fluorobenzeneboronic acid, 98%

H52598.MDEA 60.4 EUR
ALFAH52598.MD ALFAH52598.03
3-Benzylcarbamoyl-4-fluorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-22-4
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
Melting Point: 237-239°
MDL No.: MFCD08235041
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR