2-Fluor-5-(2-fluorbenzylcarbamoyl)phenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

H54510.03EA 520 EUR
H54510.03 H54510.MD
2-Fluor-5-(2-fluorbenzylcarbamoyl)phenylboronsäure ≥97%
2-Fluor-5-(2-fluorbenzylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₂BF₂NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20040208
CAS-Nummer: 1449144-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR