2-Fluoro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H54684.MDEA 85.7 EUR
ALFAH54684.MD ALFAH54684.03
2-Fluoro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20265322
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR