Magnesiumnitrid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL415111-10GEA 93.3 EUR
SIAL415111-10G
Magnesiumnitrid, Sigma-Aldrich®
Magnesiumnitrid
Formel: Mg₃N₂
Molekulargewicht: 100,93 g/mol
MDL Number: MFCD00016205
CAS-Nummer: 12057-71-5
EINECS: 235-022-1
UN-Nummer: 3178
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR