Propargit
Lieferant: EHRENSTORFER GMBH
EHERC16430000EA
78.2
EUR
EHERC16430000
Propargit
Propargit
Formel:
C₁₉H₂₆O₄S |
MDL Number:
MFCD00072448 CAS-Nummer: 2312-35-8 EINECS: 219-006-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...