(R)-(-)-2-Decanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL557986-1GEA 153 EUR
SIAL557986-1G
(R)-(-)-2-Decanol, Sigma-Aldrich®
(R)-(-)-2-Decanol
Formel: C₁₀H₂₂O
MDL Number: MFCD03427199
CAS-Nummer: 33758-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR