Ethyl-vinylsulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL445916-5MLEA 103 EUR
SIAL445916-5ML
Ethyl-vinylsulfid, Sigma-Aldrich®
Ethyl-vinylsulfid
Formel: C₄H₈S
MDL Number: MFCD00015180
CAS-Nummer: 627-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR