4-n-Octyloxybenzeneboronic acid, 97%

H52653.03EA 27.6 EUR
ALFAH52653.03 ALFAH52653.06
4-n-Octyloxybenzeneboronic acid, 97%
Purity: 97%
CAS No.: 121554-09-4
Molecular Formula: C14H23BO3
Formula Weight: 250.14
Melting Point: 88-90°
MDL No.: MFCD00982004
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR