3,5-Dibrom-p-toluidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL658367-5GEA 135 EUR
SIAL658367-5G
3,5-Dibrom-p-toluidin, Sigma-Aldrich®
3,5-Dibrom-p-toluidin
Formel: C₇H₇Br₂N
Molekulargewicht: 264,96 g/mol
MDL Number: MFCD00151806
CAS-Nummer: 13194-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR