3-Chlor-2,4-difluorbenzylamin 97%

Lieferant: Thermo Scientific
Gefahr

H26723.03EA 106 EUR
H26723.03 H26723.06
3-Chlor-2,4-difluorbenzylamin 97%
3-Chlor-2,4-difluorbenzylamin
Formel: C₇H₆ClF₂N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660335
CAS-Nummer: 886761-65-5
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR