4-Chlor-2,6-difluorbenzylamin 97%

Lieferant: Thermo Scientific

Gefahr

H26446.06EA 377 EUR
H26446.06 H26446.03
4-Chlor-2,6-difluorbenzylamin 97%
4-Chlor-2,6-difluorbenzylamin
Formel: C₇H₆ClF₂N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660350
CAS-Nummer: 518357-43-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR